Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FRA10AC1
Entrez Gene ID
118924
Gene Name
fragile site, folic acid type, rare, fra(10)(q23.3) or fra(10)(q24.2) candidate 1
Gene Aliases
C10orf4, F26C11.1-like, FRA10A
Chromosome Location
Chr.10: 93667883 - 93702572 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145246.4 NP_660289.2 6 - 7 584 135
XM_005269515.4 XP_005269572.1 7 - 8 681 135
XM_005269516.4 XP_005269573.1 6 - 7 776 135
XM_011539259.2 XP_011537561.1 6 - 7 549 135
XM_011539260.2 XP_011537562.1 7 - 8 639 135
XM_017015656.1 XP_016871145.1 6 - 7 549 135
XM_017015657.1 XP_016871146.1 6 - 7 584 135
XM_017015658.1 XP_016871147.1 7 - 8 684 135
XM_017015659.1 XP_016871148.1 6 - 7 776 135
XM_017015660.1 XP_016871149.1 7 - 8 641 135
XM_017015661.1 XP_016871150.1 7 - 8 698 135
XM_017015662.1 XP_016871151.1 6 - 7 549 135
GenBank mRNA AJ431721.1 - 6 - 7 580 135
AJ431722.1 - 6 - 7 580 135
AJ431723.1 - 6 - 7 580 135
AJ431724.1 - 6 - 7 580 135
AJ431725.1 - 6 - 7 580 135
AK090955.1 - 6 - 7 584 135
BC018007.1 - 6 - 7 453 135

View Full Product Details