Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDLIM5
Entrez Gene ID
10611
Gene Name
PDZ and LIM domain 5
Gene Aliases
ENH, ENH1, L9, LIM
Chromosome Location
Chr.4: 94451857 - 94668227 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001011513.3 NP_001011513.3 12 - 13 1548 65
NM_001256425.1 NP_001243354.1 11 - 12 1456 65
NM_001256426.1 NP_001243355.1 16 - 17 1962 65
NM_001256427.1 NP_001243356.1 13 - 14 1566 65
NM_001256428.1 NP_001243357.1 11 - 12 1723 65
NM_006457.4 NP_006448.4 12 - 13 1875 65
XM_005262693.4 XP_005262750.1 16 - 17 2259 65
XM_005262695.4 XP_005262752.1 15 - 16 2241 65
XM_005262696.4 XP_005262753.1 15 - 16 1859 65
XM_005262698.4 XP_005262755.1 13 - 14 1488 65
XM_006714066.3 XP_006714129.1 17 - 18 2274 65
XM_006714068.3 XP_006714131.1 17 - 18 1892 65
XM_006714069.3 XP_006714132.1 14 - 15 1878 65
XM_006714070.3 XP_006714133.1 17 - 18 1875 65
XM_011531543.2 XP_011529845.1 16 - 17 1857 65
XM_017007657.1 XP_016863146.1 13 - 14 1863 65
XM_017007658.1 XP_016863147.1 14 - 15 1496 65
GenBank mRNA AB209531.1 - 8 - 9 1587 65
AF061258.1 - 12 - 13 1777 65
AK027217.1 - 2 - 3 1794 65
AK291624.1 - 12 - 13 1797 65
AK296980.1 - 11 - 12 1637 65
AK304125.1 - 13 - 14 1488 65
AL832157.1 - 12 - 13 1450 65
AL832686.1 - 16 - 17 1866 65
AY598328.1 - 11 - 12 1694 65
BC008741.2 - 12 - 13 1814 65
BC026029.2 - 10 - 11 1318 65
BF954250.1 - 1 - 2 65
BT007328.1 - 11 - 12 1694 65

View Full Product Details