Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SMAD9
Entrez Gene ID
4093
Gene Name
SMAD family member 9
Gene Aliases
MADH6, MADH9, PPH2, SMAD8, SMAD8/9, SMAD8A, SMAD8B
Chromosome Location
Chr.13: 36844831 - 36920721 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127217.2 NP_001120689.1 3 - 4 1017 68
XM_006719827.3 XP_006719890.1 3 - 4 947 68
GenBank mRNA AK311686.1 - 1 - 2 52 68
AK314920.1 - 2 - 3 726 68
BC104760.1 - 2 - 3 769 68
BC104762.1 - 2 - 3 769 68
BC143240.1 - 2 - 3 769 68
D83760.1 - 2 - 3 729 68

View Full Product Details