Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MSMO1
Entrez Gene ID
6307
Gene Name
methylsterol monooxygenase 1
Gene Aliases
DESP4, ERG25, MCCPD, SC4MOL
Chromosome Location
Chr.4: 165327666 - 165343164 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005263176.2 XP_005263233.1 1 - 2 222 91

View Full Product Details