The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000016.5 | NP_000007.1 | 10 - 11 | 1380 | 122 |
NM_001127328.2 | NP_001120800.1 | 10 - 11 | 1392 | 122 | |
NM_001286042.1 | NP_001272971.1 | 9 - 10 | 1292 | 122 | |
NM_001286043.1 | NP_001272972.1 | 11 - 12 | 1479 | 122 | |
NM_001286044.1 | NP_001272973.1 | 7 - 8 | 1110 | 122 | |
GenBank mRNA | AF251043.1 | - | 10 - 11 | 959 | 122 |
AK296045.1 | - | 7 - 8 | 690 | 122 | |
AK301005.1 | - | 7 - 8 | 848 | 122 | |
AK301039.1 | - | 10 - 11 | 1036 | 122 | |
AK301717.1 | - | 9 - 10 | 884 | 122 | |
AK312629.1 | - | 10 - 11 | 970 | 122 | |
AK315946.1 | - | 9 - 10 | 933 | 122 | |
BC005377.1 | - | 10 - 11 | 974 | 122 | |
BX647734.1 | - | 11 - 12 | 1118 | 122 | |
KU177876.1 | - | 10 - 11 | 939 | 122 | |
KU177877.1 | - | 9 - 10 | 851 | 122 | |
M16827.1 | - | 10 - 11 | 957 | 122 |