The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000016.5 | NP_000007.1 | 2 - 3 | 560 | 82 |
NM_001127328.2 | NP_001120800.1 | 2 - 3 | 572 | 82 | |
NM_001286043.1 | NP_001272972.1 | 2 - 3 | 560 | 82 | |
GenBank mRNA | AF251043.1 | - | 2 - 3 | 139 | 82 |
AK301039.1 | - | 2 - 3 | 201 | 82 | |
AK311022.1 | - | 2 - 3 | 142 | 82 | |
AK312629.1 | - | 2 - 3 | 150 | 82 | |
BC005377.1 | - | 2 - 3 | 154 | 82 | |
BX647734.1 | - | 2 - 3 | 199 | 82 | |
KU177876.1 | - | 2 - 3 | 119 | 82 | |
M16827.1 | - | 2 - 3 | 137 | 82 |