The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000016.5 | NP_000007.1 | 9 - 10 | 1289 | 143 |
NM_001127328.2 | NP_001120800.1 | 9 - 10 | 1301 | 143 | |
NM_001286042.1 | NP_001272971.1 | 8 - 9 | 1201 | 143 | |
NM_001286043.1 | NP_001272972.1 | 10 - 11 | 1388 | 143 | |
NM_001286044.1 | NP_001272973.1 | 6 - 7 | 1019 | 143 | |
GenBank mRNA | AF251043.1 | - | 9 - 10 | 868 | 143 |
AK296045.1 | - | 6 - 7 | 599 | 143 | |
AK301005.1 | - | 6 - 7 | 757 | 143 | |
AK301039.1 | - | 9 - 10 | 945 | 143 | |
AK301717.1 | - | 8 - 9 | 793 | 143 | |
AK312629.1 | - | 9 - 10 | 879 | 143 | |
AK315946.1 | - | 8 - 9 | 842 | 143 | |
BC005377.1 | - | 9 - 10 | 883 | 143 | |
BX647734.1 | - | 10 - 11 | 1027 | 143 | |
KU177876.1 | - | 9 - 10 | 848 | 143 | |
KU177877.1 | - | 8 - 9 | 760 | 143 | |
M16827.1 | - | 9 - 10 | 866 | 143 |