The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030768.2 | NP_110395.1 | 10 - 11 | 1082 | 72 |
XM_005246106.1 | XP_005246163.1 | 10 - 11 | 1065 | 72 | |
XM_006712784.1 | XP_006712847.1 | 10 - 11 | 1117 | 72 | |
XM_011511946.2 | XP_011510248.1 | 9 - 10 | 942 | 72 | |
XM_017005056.1 | XP_016860545.1 | 10 - 11 | 941 | 72 | |
XM_017005057.1 | XP_016860546.1 | 10 - 11 | 1455 | 72 | |
XM_017005058.1 | XP_016860547.1 | 9 - 10 | 980 | 72 | |
GenBank mRNA | AK001043.1 | - | 10 - 11 | 1094 | 72 |
AK055417.1 | - | 5 - 6 | 2300 | 72 | |
AL136850.1 | - | 10 - 11 | 1007 | 72 | |
AY024365.1 | - | 10 - 11 | 1053 | 72 | |
BC006576.2 | - | 10 - 11 | 1082 | 72 | |
BC035746.1 | - | 9 - 10 | 1477 | 72 | |
CR533533.1 | - | 10 - 11 | 952 | 72 | |
FJ491380.1 | - | 10 - 11 | 975 | 72 | |
HQ658059.1 | - | 9 - 10 | 975 | 72 | |
HQ658060.1 | - | 7 - 8 | 728 | 72 |