The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030768.2 | NP_110395.1 | 3 - 4 | 309 | 126 |
XM_011511946.2 | XP_011510248.1 | 3 - 4 | 306 | 126 | |
XM_017005056.1 | XP_016860545.1 | 3 - 4 | 178 | 126 | |
GenBank mRNA | AK001043.1 | - | 3 - 4 | 321 | 126 |
AL136850.1 | - | 3 - 4 | 234 | 126 | |
AY024365.1 | - | 3 - 4 | 280 | 126 | |
BC006576.2 | - | 3 - 4 | 309 | 126 | |
CR533533.1 | - | 3 - 4 | 179 | 126 | |
FJ491381.1 | - | 3 - 4 | 179 | 126 | |
HQ658059.1 | - | 3 - 4 | 309 | 126 |