Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM54
Entrez Gene ID
57159
Gene Name
tripartite motif containing 54
Gene Aliases
MURF, MURF-3, RNF30, muRF3
Chromosome Location
Chr.2: 27282429 - 27307439 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032546.3 NP_115935.3 7 - 8 1298 54
NM_187841.2 NP_912730.2 6 - 7 1172 54
GenBank mRNA AJ291714.1 - 7 - 8 995 54
AK300699.1 - 7 - 8 1144 54
BC141807.1 - 6 - 7 944 54

View Full Product Details