The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032023.3 | NP_114412.2 | 3 - 4 | 266 | 65 |
XM_005271833.4 | XP_005271890.1 | 3 - 4 | 235 | 65 | |
XM_005271835.4 | XP_005271892.1 | 3 - 4 | 235 | 65 | |
XM_006718013.3 | XP_006718076.1 | 3 - 4 | 235 | 65 | |
XM_006718014.3 | XP_006718077.1 | 2 - 3 | 1780 | 65 | |
XM_017016754.1 | XP_016872243.1 | 2 - 3 | 1780 | 65 | |
XM_017016755.1 | XP_016872244.1 | 2 - 3 | 1780 | 65 | |
XM_017016756.1 | XP_016872245.1 | 3 - 4 | 235 | 65 | |
XM_017016757.1 | XP_016872246.1 | 2 - 3 | 1780 | 65 | |
GenBank mRNA | AB209446.1 | - | 2 - 3 | 2138 | 65 |
AF260335.1 | - | 3 - 4 | 230 | 65 | |
AK055763.1 | - | 1 - 2 | 1274 | 65 | |
AY216713.1 | - | 3 - 4 | 266 | 65 | |
AY216716.1 | - | 3 - 4 | 266 | 65 | |
BC032593.1 | - | 3 - 4 | 248 | 65 | |
BI601296.1 | - | 3 - 4 | 269 | 65 |