Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DSG2
Entrez Gene ID
1829
Gene Name
desmoglein 2
Gene Aliases
CDHF5, HDGC
Chromosome Location
Chr.18: 31498004 - 31549008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001943.4 NP_001934.2 13 - 14 2252 59
GenBank mRNA AK308408.1 - 13 - 14 2009 59
BC099655.3 - 13 - 14 2015 59
BC099656.3 - 13 - 14 2015 59
BC099657.3 - 13 - 14 2015 59
Z26317.1 - 13 - 14 2012 59

View Full Product Details