Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DSG2
Entrez Gene ID
1829
Gene Name
desmoglein 2
Gene Aliases
CDHF5, HDGC
Chromosome Location
Chr.18: 31498004 - 31549008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001943.4 NP_001934.2 7 - 8 1077 143
GenBank mRNA AK308408.1 - 7 - 8 834 143
BC099655.3 - 7 - 8 840 143
BC099656.3 - 7 - 8 840 143
BC099657.3 - 7 - 8 840 143
Z26317.1 - 7 - 8 837 143

View Full Product Details