Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PHACTR3
Entrez Gene ID
116154
Gene Name
phosphatase and actin regulator 3
Gene Aliases
C20orf101, H17739, PPP1R123, SCAPIN1, SCAPININ
Chromosome Location
Chr.20: 59577497 - 59849829 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_017027627.1 XP_016883116.1 1 - 2 260 93
XM_017027629.1 XP_016883118.1 1 - 2 261 93

View Full Product Details