The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145548.1 | NP_001139020.1 | 8 - 9 | 1212 | 65 |
NM_017740.2 | NP_060210.2 | 7 - 8 | 1101 | 65 | |
XM_017023434.1 | XP_016878923.1 | 5 - 6 | 637 | 65 | |
XM_017023435.1 | XP_016878924.1 | 5 - 6 | 497 | 65 | |
XM_017023436.1 | XP_016878925.1 | 5 - 6 | 612 | 65 | |
GenBank mRNA | AK000286.1 | - | 7 - 8 | 1101 | 65 |
AK001654.1 | - | 5 - 6 | 487 | 65 | |
AK225768.1 | - | 7 - 8 | 1053 | 65 | |
BC017702.1 | - | 5 - 6 | 462 | 65 | |
BC018772.1 | - | 8 - 9 | 1199 | 65 |