The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001290229.1 | NP_001277158.1 | 4 - 5 | 565 | 137 |
NM_004866.5 | NP_004857.4 | 5 - 6 | 643 | 137 | |
XM_011543727.2 | XP_011542029.1 | 5 - 6 | 643 | 137 | |
GenBank mRNA | AF005037.1 | - | 5 - 6 | 517 | 137 |
AF038966.1 | - | 5 - 6 | 564 | 137 | |
AK225172.1 | - | 5 - 6 | 532 | 137 | |
AK290225.1 | - | 5 - 6 | 493 | 137 | |
AK304452.1 | - | 4 - 5 | 457 | 137 | |
BC015065.1 | - | 5 - 6 | 536 | 137 | |
BC034048.1 | - | 5 - 6 | 497 | 137 | |
CR542286.1 | - | 5 - 6 | 474 | 137 |