Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIFC3
Entrez Gene ID
3801
Gene Name
kinesin family member C3
Gene Aliases
-
Chromosome Location
Chr.16: 57758217 - 57863053 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130099.1 NP_001123571.1 4 - 5 663 91
NM_001130100.1 NP_001123572.1 5 - 6 719 91
NM_001318710.1 NP_001305639.1 5 - 6 600 91
NM_001318711.1 NP_001305640.1 5 - 6 761 91
NM_001318712.1 NP_001305641.1 3 - 4 336 91
NM_001318713.1 NP_001305642.1 3 - 4 738 91
NM_001318714.1 NP_001305643.1 6 - 7 1108 91
NM_001318715.1 NP_001305644.1 5 - 6 697 91
NM_005550.3 NP_005541.3 5 - 6 719 91
NR_134678.1 5 - 6 903 91
XM_005255937.1 XP_005255994.1 7 - 8 1228 91
XM_005255944.1 XP_005256001.1 5 - 6 582 91
XM_006721188.1 XP_006721251.1 8 - 9 1306 91
XM_011523075.1 XP_011521377.1 5 - 6 886 91
XM_011523076.1 XP_011521378.1 5 - 6 886 91
XM_011523077.1 XP_011521379.1 5 - 6 685 91
XM_011523078.1 XP_011521380.1 5 - 6 488 91
XM_011523079.1 XP_011521381.1 5 - 6 823 91
XM_017023219.1 XP_016878708.1 8 - 9 1275 91
XM_017023220.1 XP_016878709.1 8 - 9 1452 91
XM_017023221.1 XP_016878710.1 7 - 8 1228 91
XM_017023222.1 XP_016878711.1 9 - 10 1470 91
XM_017023223.1 XP_016878712.1 5 - 6 603 91
XM_017023224.1 XP_016878713.1 7 - 8 1137 91
XM_017023225.1 XP_016878714.1 3 - 4 1707 91
XM_017023226.1 XP_016878715.1 6 - 7 980 91
XM_017023227.1 XP_016878716.1 6 - 7 980 91
GenBank mRNA AB209290.1 - 5 - 6 654 91
AF004426.1 - 1 - 2 105 91
AK091891.1 - - 1649 91
AK291737.1 - 4 - 5 558 91
AK296995.1 - 5 - 6 599 91
AK299431.1 - 5 - 6 661 91
AK302743.1 - 3 - 4 336 91
AK303021.1 - 3 - 4 666 91
AK304132.1 - 5 - 6 707 91
AK307846.1 - 5 - 6 677 91
BC001211.1 - 5 - 6 688 91
BC034234.1 - 6 - 7 1048 91
BC041132.1 - 5 - 6 549 91
BC047051.1 - 5 - 6 847 91

View Full Product Details