The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024927.4 | NP_079203.3 | 11 - 12 | 2478 | 72 |
XM_017025113.1 | XP_016880602.1 | 12 - 13 | 2449 | 72 | |
XM_017025114.1 | XP_016880603.1 | 12 - 13 | 2440 | 72 | |
XM_017025115.1 | XP_016880604.1 | 12 - 13 | 2599 | 72 | |
XM_017025116.1 | XP_016880605.1 | 11 - 12 | 2314 | 72 | |
XM_017025117.1 | XP_016880606.1 | 12 - 13 | 2269 | 72 | |
XM_017025118.1 | XP_016880607.1 | 11 - 12 | 2455 | 72 | |
GenBank mRNA | AL137584.1 | - | 1 - 2 | 237 | 72 |
BC052978.1 | - | 11 - 12 | 2399 | 72 |