The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014825.2 | NP_055640.2 | 17 - 18 | 2337 | 68 |
GenBank mRNA | AB011111.2 | - | 12 - 13 | 1571 | 68 |
AF231919.1 | - | 17 - 18 | 2175 | 68 |