Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS36
Entrez Gene ID
51028
Gene Name
vacuolar protein sorting 36 homolog
Gene Aliases
C13orf9, CGI-145, EAP45
Chromosome Location
Chr.13: 52412602 - 52450678 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282168.1 NP_001269097.1 13 - 14 1125 76
NM_001282169.1 NP_001269098.1 13 - 14 1286 76
NM_016075.3 NP_057159.2 13 - 14 1152 76
GenBank mRNA AB208956.1 - 7 - 8 3826 76
AF151903.1 - 13 - 14 1073 76
AK023182.1 - 11 - 12 869 76
AK289740.1 - 13 - 14 1095 76
BC037279.1 - 13 - 14 1101 76
BC050439.1 - 13 - 14 1075 76
BX647847.1 - 13 - 14 1093 76
CR933653.1 - 13 - 14 1240 76

View Full Product Details