The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018676.3 | NP_061146.1 | 1 - 2 | 467 | 138 |
NM_199263.2 | NP_954872.1 | 1 - 2 | 467 | 138 | |
GenBank mRNA | AK096289.1 | - | 1 - 2 | 467 | 138 |
AK315087.1 | - | 1 - 2 | 116 | 138 | |
AY358149.1 | - | 1 - 2 | 130 | 138 |