Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STMN4
Entrez Gene ID
81551
Gene Name
stathmin 4
Gene Aliases
RB3
Chromosome Location
Chr.8: 27235323 - 27258865 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001283053.1 NP_001269982.1 4 - 5 359 77
NM_030795.3 NP_110422.2 4 - 5 359 77
XM_005273652.4 XP_005273709.1 4 - 5 785 77
GenBank mRNA AK294023.1 - 4 - 5 324 77
AK297415.1 - 4 - 5 340 77
BC011520.1 - 4 - 5 306 77

View Full Product Details