Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTDSP2
Entrez Gene ID
10106
Gene Name
CTD small phosphatase 2
Gene Aliases
OS4, PSR2, SCP2
Chromosome Location
Chr.12: 57819927 - 57846964 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005730.3 NP_005721.3 2 - 3 742 71
XM_005268556.1 XP_005268613.1 2 - 3 502 71
GenBank mRNA AF000152.1 - 2 - 3 518 71
AF022231.1 - 2 - 3 459 71
AK226076.1 - 2 - 3 498 71
AK291289.1 - 2 - 3 489 71
AK310873.1 - 2 - 3 244 71
AK312413.1 - 2 - 3 501 71
AY279531.1 - 2 - 3 213 71
BC018922.2 - 2 - 3 474 71
BC065920.1 - 2 - 3 457 71

View Full Product Details