Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CTDSP2
Entrez Gene ID
10106
Gene Name
CTD small phosphatase 2
Gene Aliases
OS4, PSR2, SCP2
Chromosome Location
Chr.12: 57819927 - 57846964 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_005730.3 NP_005721.3 3 - 4 777 85
GenBank mRNA AF000152.1 - 3 - 4 553 85
AF022231.1 - 3 - 4 494 85
AK226076.1 - 3 - 4 533 85
AK291289.1 - 3 - 4 524 85
AK310873.1 - 3 - 4 279 85
AK312413.1 - 3 - 4 536 85
AY279531.1 - 3 - 4 248 85
BC018922.2 - 3 - 4 509 85
BC065920.1 - 3 - 4 492 85

View Full Product Details