Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DZIP1
Entrez Gene ID
22873
Gene Name
DAZ interacting zinc finger protein 1
Gene Aliases
DZIP, DZIPt1
Chromosome Location
Chr.13: 95578202 - 95644706 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014934.4 NP_055749.1 1 - 2 329 104
NM_198968.3 NP_945319.1 1 - 2 329 104
XM_005254032.1 XP_005254089.1 1 - 2 329 104
XM_006719916.1 XP_006719979.1 1 - 2 329 104
XM_006719917.2 XP_006719980.1 1 - 2 329 104
XM_006719918.1 XP_006719981.1 1 - 2 329 104
XM_006719920.1 XP_006719983.1 1 - 2 329 104
XM_017020444.1 XP_016875933.1 1 - 2 329 104
XM_017020445.1 XP_016875934.1 1 - 2 329 104
XM_017020446.1 XP_016875935.1 1 - 2 329 104
GenBank mRNA AB023213.1 - 1 - 2 313 104
BC041804.1 - 1 - 2 329 104
DA782750.1 - 1 - 2 330 104
DR001764.1 - 1 - 2 249 104

View Full Product Details