The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282683.1 | NP_001269612.1 | 5 - 6 | 457 | 75 |
NM_024735.4 | NP_079011.3 | 4 - 5 | 709 | 75 | |
GenBank mRNA | AF318348.1 | - | 3 - 4 | 909 | 75 |
AF428140.1 | - | 4 - 5 | 673 | 75 | |
AL117444.1 | - | 2 - 3 | 217 | 75 | |
AY736035.1 | - | 5 - 6 | 660 | 75 | |
BC012748.1 | - | 4 - 5 | 683 | 75 | |
BG718342.1 | - | 5 - 6 | 75 |