The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012227.3 | NP_036359.3 | 5 - 6 | 781 | 77 |
XM_006724868.3 | XP_006724931.1 | 6 - 7 | 681 | 77 | |
XM_011545637.2 | XP_011543939.1 | 4 - 5 | 386 | 77 | |
GenBank mRNA | AK024630.1 | - | 5 - 6 | 720 | 77 |
AK223412.1 | - | 5 - 6 | 726 | 77 | |
AK296003.1 | - | 5 - 6 | 442 | 77 | |
AK298260.1 | - | 5 - 6 | 528 | 77 | |
BC014636.1 | - | 5 - 6 | 454 | 77 | |
BT007360.1 | - | 4 - 5 | 410 | 77 | |
Y14391.2 | - | 5 - 6 | 739 | 77 |