Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TRIM46
Entrez Gene ID
80128
Gene Name
tripartite motif containing 46
Gene Aliases
GENEY, TRIFIC
Chromosome Location
Chr.1: 155173714 - 155184971 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256599.1 NP_001243528.1 4 - 5 1001 71
NM_001256600.1 NP_001243529.1 5 - 6 1032 71
NM_001256601.1 NP_001243530.1 4 - 5 832 71
NM_001282378.1 NP_001269307.1 3 - 4 729 71
NM_001282379.1 NP_001269308.1 4 - 5 991 71
NM_025058.4 NP_079334.3 4 - 5 991 71
NR_046327.1 4 - 5 991 71
NR_046329.1 5 - 6 1050 71
NR_104150.1 4 - 5 991 71
XM_011510002.1 XP_011508304.1 5 - 6 934 71
XM_017002393.1 XP_016857882.1 5 - 6 1105 71
GenBank mRNA AK026882.1 - 4 - 5 902 71
AK096158.1 - 4 - 5 879 71
AK128139.1 - 5 - 6 1050 71
AK225075.1 - 4 - 5 902 71
AK296300.1 - 5 - 6 1028 71
AK304542.1 - 4 - 5 835 71
AK308075.1 - 4 - 5 796 71
AK309699.1 - 4 - 5 832 71
AK310429.1 - 5 - 6 1137 71
AY251386.1 - 4 - 5 811 71
BC069416.1 - 4 - 5 887 71
BC069568.1 - 4 - 5 887 71
BC126372.1 - 4 - 5 908 71
BC143828.1 - 4 - 5 908 71
BX283909.1 - 1 - 2 97 71
DB262381.1 - 2 - 3 481 71

View Full Product Details