Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SECISBP2L
Entrez Gene ID
9728
Gene Name
SECIS binding protein 2 like
Gene Aliases
SBP2L, SLAN
Chromosome Location
Chr.15: 48988638 - 49046563 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001193489.1 NP_001180418.1 5 - 6 1152 104
NM_014701.3 NP_055516.2 5 - 6 1152 104
GenBank mRNA AK307480.1 - 5 - 6 835 104
BC033001.2 - 5 - 6 1087 104
D87445.3 - 5 - 6 1022 104

View Full Product Details