The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001193489.1 | NP_001180418.1 | 6 - 7 | 1185 | 86 |
NM_014701.3 | NP_055516.2 | 6 - 7 | 1185 | 86 | |
GenBank mRNA | AK307480.1 | - | 6 - 7 | 868 | 86 |
AK308183.1 | - | 6 - 7 | 820 | 86 | |
BC033001.2 | - | 6 - 7 | 1120 | 86 | |
D87445.3 | - | 6 - 7 | 1055 | 86 |