The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_104371.3 | 11 - 12 | 1354 | 79 | |
GenBank mRNA | AY341345.1 | - | 10 - 11 | 1102 | 79 |
AY353056.1 | - | 1 - 2 | 59 | 79 | |
AY353057.1 | - | 12 - 13 | 1309 | 79 | |
AY353058.1 | - | 11 - 12 | 1225 | 79 | |
AY353060.1 | - | 11 - 12 | 1216 | 79 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001018100.4 | NP_001018110.1 | 12 - 13 | 1438 | 79 |
NM_152451.7 | NP_689664.3 | 11 - 12 | 1354 | 79 | |
GenBank mRNA | AK055535.1 | - | 12 - 13 | 1401 | 79 |
AL833028.1 | - | 7 - 8 | 681 | 79 | |
AL833142.1 | - | 10 - 11 | 1135 | 79 | |
AY207459.1 | - | 12 - 13 | 1309 | 79 | |
AY237639.1 | - | 11 - 12 | 1225 | 79 | |
BC101645.1 | - | 12 - 13 | 1438 | 79 | |
BC112148.1 | - | 12 - 13 | 1436 | 79 | |
FJ970029.1 | - | 12 - 13 | 1314 | 79 |