The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_073573.1 | 11 - 12 | 2453 | 73 | |
NR_073574.1 | 11 - 12 | 1825 | 73 | ||
GenBank mRNA | AK096767.1 | - | 11 - 12 | 1825 | 73 |
BC016062.1 | - | 11 - 12 | 2056 | 73 |