Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAMP3
Entrez Gene ID
10268
Gene Name
receptor (G protein-coupled) activity modifying protein 3
Gene Aliases
-
Chromosome Location
Chr.7: 45157768 - 45184250 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006715631.3 XP_006715694.1 1 - 2 114 68

View Full Product Details