Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM172A
Entrez Gene ID
83989
Gene Name
family with sequence similarity 172 member A
Gene Aliases
C5orf21
Chromosome Location
Chr.5: 93615130 - 94111699 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163417.1 NP_001156889.1 6 - 7 866 111
NM_001163418.1 NP_001156890.1 5 - 6 674 111
NM_032042.5 NP_114431.2 7 - 8 990 111
NR_028080.1 5 - 6 697 111
XM_005272103.3 XP_005272160.1 7 - 8 956 111
XM_005272105.3 XP_005272162.1 6 - 7 763 111
XM_005272108.3 XP_005272165.1 6 - 7 884 111
XM_005272109.4 XP_005272166.1 7 - 8 956 111
XM_006714717.3 XP_006714780.1 7 - 8 982 111
XM_006714718.3 XP_006714781.1 5 - 6 1596 111
XM_006714719.3 XP_006714782.1 5 - 6 745 111
XM_011543668.2 XP_011541970.1 7 - 8 956 111
XM_011543670.2 XP_011541972.1 7 - 8 956 111
XM_011543671.2 XP_011541973.1 7 - 8 956 111
XM_011543672.2 XP_011541974.1 7 - 8 886 111
XM_011543673.2 XP_011541975.1 6 - 7 785 111
XM_011543674.2 XP_011541976.1 7 - 8 956 111
XM_017009952.1 XP_016865441.1 7 - 8 1278 111
XM_017009953.1 XP_016865442.1 7 - 8 1276 111
XM_017009954.1 XP_016865443.1 7 - 8 956 111
XM_017009955.1 XP_016865444.1 6 - 7 1694 111
XM_017009956.1 XP_016865445.1 6 - 7 1008 111
XM_017009957.1 XP_016865446.1 5 - 6 962 111
XM_017009958.1 XP_016865447.1 5 - 6 757 111
XM_017009959.1 XP_016865448.1 6 - 7 1035 111
XM_017009960.1 XP_016865449.1 4 - 5 539 111
XM_017009961.1 XP_016865450.1 6 - 7 708 111
GenBank mRNA AK295127.1 - 6 - 7 889 111
AK295879.1 - 5 - 6 623 111
AK296986.1 - 6 - 7 843 111
AK297471.1 - 4 - 5 459 111
AK312930.1 - 7 - 8 895 111
BC020584.2 - 6 - 7 792 111

View Full Product Details