The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032042.5 | NP_114431.2 | 2 - 3 | 309 | 108 |
NR_028080.1 | 2 - 3 | 309 | 108 | ||
XM_005272103.3 | XP_005272160.1 | 2 - 3 | 275 | 108 | |
XM_005272105.3 | XP_005272162.1 | 2 - 3 | 274 | 108 | |
XM_005272106.3 | XP_005272163.1 | 2 - 3 | 275 | 108 | |
XM_005272108.3 | XP_005272165.1 | 2 - 3 | 304 | 108 | |
XM_005272109.4 | XP_005272166.1 | 2 - 3 | 275 | 108 | |
XM_005272111.4 | XP_005272168.1 | 2 - 3 | 275 | 108 | |
XM_006714717.3 | XP_006714780.1 | 2 - 3 | 301 | 108 | |
XM_011543668.2 | XP_011541970.1 | 2 - 3 | 275 | 108 | |
XM_011543670.2 | XP_011541972.1 | 2 - 3 | 275 | 108 | |
XM_011543671.2 | XP_011541973.1 | 2 - 3 | 275 | 108 | |
XM_011543674.2 | XP_011541976.1 | 2 - 3 | 275 | 108 | |
XM_017009954.1 | XP_016865443.1 | 2 - 3 | 275 | 108 | |
XM_017009961.1 | XP_016865450.1 | 2 - 3 | 219 | 108 | |
GenBank mRNA | AK295127.1 | - | 2 - 3 | 309 | 108 |
AK297471.1 | - | 1 - 2 | 71 | 108 | |
AK312930.1 | - | 2 - 3 | 214 | 108 | |
AL136630.1 | - | 2 - 3 | 148 | 108 | |
BC020584.2 | - | 1 - 2 | 111 | 108 | |
DC324758.1 | - | 2 - 3 | 309 | 108 | |
DC374283.1 | - | 2 - 3 | 294 | 108 |