The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014321.3 | NP_055136.1 | 1 - 2 | 128 | 153 |
XM_011522978.2 | XP_011521280.1 | 1 - 2 | 118 | 153 | |
GenBank mRNA | AF139658.1 | - | 1 - 2 | 116 | 153 |
AK024019.1 | - | 1 - 2 | 122 | 153 |