The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002037.5 | NP_002028.1 | 4 - 5 | 848 | 89 |
NM_153047.3 | NP_694592.1 | 1 - 2 | 252 | 89 | |
NM_153048.3 | NP_694593.1 | 2 - 3 | 344 | 89 | |
XM_005266890.3 | XP_005266947.1 | 4 - 5 | 429 | 89 | |
XM_005266892.3 | XP_005266949.1 | 4 - 5 | 429 | 89 | |
XM_017010650.1 | XP_016866139.1 | 3 - 4 | 586 | 89 | |
XM_017010651.1 | XP_016866140.1 | 3 - 4 | 490 | 89 | |
XM_017010652.1 | XP_016866141.1 | 2 - 3 | 500 | 89 | |
XM_017010653.1 | XP_016866142.1 | 2 - 3 | 344 | 89 | |
XM_017010654.1 | XP_016866143.1 | 2 - 3 | 325 | 89 | |
XM_017010655.1 | XP_016866144.1 | 4 - 5 | 429 | 89 | |
GenBank mRNA | AB451293.1 | - | 1 - 2 | 241 | 89 |
AB451426.1 | - | 1 - 2 | 241 | 89 | |
AK056699.1 | - | 3 - 4 | 581 | 89 | |
BC032496.1 | - | 2 - 3 | 295 | 89 | |
CX781584.1 | - | 3 - 4 | 505 | 89 | |
CX872011.1 | - | 2 - 3 | 282 | 89 | |
DA815564.1 | - | 2 - 3 | 345 | 89 | |
DA922886.1 | - | 2 - 3 | 339 | 89 | |
M14333.1 | - | 4 - 5 | 820 | 89 | |
M14676.1 | - | 4 - 5 | 611 | 89 | |
S74774.1 | - | 1 - 2 | 241 | 89 |