Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BUD13
Entrez Gene ID
84811
Gene Name
BUD13 homolog
Gene Aliases
Cwc26, fSAP71
Chromosome Location
Chr.11: 116748170 - 116772998 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159736.1 NP_001153208.1 8 - 9 1320 123
NM_032725.3 NP_116114.1 8 - 9 1722 123
XM_011543035.2 XP_011541337.1 8 - 9 2296 123
GenBank mRNA AK057832.1 - 8 - 9 1309 123
AK225164.1 - 8 - 9 1309 123
AK289635.1 - 8 - 9 1711 123
AK291111.1 - 8 - 9 1711 123
BC006350.2 - 8 - 9 1712 123

View Full Product Details