The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_002427.3 | NP_002418.1 | 1 - 2 | 149 | 88 |
GenBank mRNA | AK222748.1 | - | 1 - 2 | 149 | 88 |
AK292211.1 | - | 1 - 2 | 149 | 88 | |
AK315341.1 | - | 1 - 2 | 147 | 88 | |
AY256443.1 | - | 1 - 2 | 120 | 88 | |
AY256444.1 | - | 1 - 2 | 120 | 88 | |
BC067522.1 | - | 1 - 2 | 128 | 88 | |
BC067523.1 | - | 1 - 2 | 128 | 88 | |
BC074807.2 | - | 1 - 2 | 139 | 88 | |
BC074808.2 | - | 1 - 2 | 139 | 88 | |
X75308.1 | - | 1 - 2 | 124 | 88 | |
X81334.1 | - | 1 - 2 | 147 | 88 |