The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000520.5 | NP_000511.2 | 2 - 3 | 849 | 72 |
NM_001318825.1 | NP_001305754.1 | 2 - 3 | 882 | 72 | |
NR_134869.1 | 2 - 3 | 849 | 72 | ||
GenBank mRNA | AK222502.1 | - | 2 - 3 | 362 | 72 |
AK296528.1 | - | 2 - 3 | 220 | 72 | |
AK297678.1 | - | 3 - 4 | 439 | 72 | |
AK301000.1 | - | 2 - 3 | 402 | 72 | |
AK301138.1 | - | 2 - 3 | 379 | 72 | |
AK307770.1 | - | 2 - 3 | 369 | 72 | |
AK310154.1 | - | 2 - 3 | 379 | 72 | |
BC018927.2 | - | 2 - 3 | 359 | 72 | |
BC084537.1 | - | 2 - 3 | 355 | 72 | |
BP356349.1 | - | 2 - 3 | 438 | 72 | |
BX537989.1 | - | 2 - 3 | 3855 | 72 | |
CR627386.1 | - | 2 - 3 | 849 | 72 | |
KU178109.1 | - | 2 - 3 | 348 | 72 | |
KU178110.1 | - | 2 - 3 | 348 | 72 | |
KU178111.1 | - | 2 - 3 | 348 | 72 | |
KU178112.1 | - | 2 - 3 | 348 | 72 | |
M13520.1 | - | 2 - 3 | 232 | 72 |