The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006240.2 | NP_006231.2 | 2 - 3 | 256 | 95 |
NM_152224.1 | NP_689410.1 | 2 - 3 | 256 | 95 | |
NM_152226.1 | NP_689412.1 | 2 - 3 | 256 | 95 | |
XM_005274553.2 | XP_005274610.1 | 4 - 5 | 583 | 95 | |
XM_017029612.1 | XP_016885101.1 | 4 - 5 | 583 | 95 | |
GenBank mRNA | BC036026.1 | - | 2 - 3 | 255 | 95 |
X97867.1 | - | 2 - 3 | 258 | 95 |