Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC10
Entrez Gene ID
54431
Gene Name
DnaJ heat shock protein family (Hsp40) member C10
Gene Aliases
ERdj5, JPDI, MTHr, PDIA19
Chromosome Location
Chr.2: 182716041 - 182780025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271581.1 NP_001258510.1 2 - 3 492 102
NM_018981.2 NP_061854.1 2 - 3 492 102
NR_073365.1 2 - 3 492 102
NR_073366.1 2 - 3 492 102
NR_073367.1 2 - 3 492 102
GenBank mRNA AF038503.1 - 2 - 3 203 102
AF490904.1 - 2 - 3 240 102
AK027647.1 - 2 - 3 261 102
AK027696.1 - 2 - 3 240 102
AL832632.2 - 2 - 3 263 102
AY358577.1 - 2 - 3 249 102
BC070034.1 - 2 - 3 237 102
BC117299.1 - 2 - 3 207 102
BC126168.1 - 2 - 3 207 102
DA106532.1 - 2 - 3 492 102

View Full Product Details