Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAJC10
Entrez Gene ID
54431
Gene Name
DnaJ heat shock protein family (Hsp40) member C10
Gene Aliases
ERdj5, JPDI, MTHr, PDIA19
Chromosome Location
Chr.2: 182716041 - 182780025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271581.1 NP_001258510.1 8 - 9 1372 83
NM_018981.2 NP_061854.1 8 - 9 1372 83
NR_073365.1 8 - 9 1372 83
NR_073366.1 8 - 9 1372 83
NR_073367.1 8 - 9 1372 83
GenBank mRNA AF038503.1 - 8 - 9 1083 83
AF490904.1 - 8 - 9 1120 83
AK027450.1 - 6 - 7 713 83
AK027696.1 - 8 - 9 1120 83
AK027701.2 - 5 - 6 494 83
AK298138.1 - 4 - 5 357 83
AL832267.1 - 1 - 2 72 83
AL832646.3 - 6 - 7 724 83
AY089971.1 - 6 - 7 726 83
AY358577.1 - 8 - 9 1129 83
BC070034.1 - 8 - 9 1117 83
BC107425.1 - 6 - 7 749 83
BC117299.1 - 8 - 9 1087 83
BC126168.1 - 8 - 9 1087 83

View Full Product Details