The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291494.1 | NP_001278423.1 | 9 - 10 | 1463 | 148 |
NM_003638.2 | NP_003629.2 | 10 - 11 | 1508 | 148 | |
XM_011519752.2 | XP_011518054.1 | 10 - 11 | 1502 | 148 | |
GenBank mRNA | AK297740.1 | - | 9 - 10 | 975 | 148 |
AK308240.1 | - | 10 - 11 | 1025 | 148 | |
L36531.1 | - | 10 - 11 | 830 | 148 |