The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291494.1 | NP_001278423.1 | 13 - 14 | 1966 | 67 |
NM_003638.2 | NP_003629.2 | 14 - 15 | 2011 | 67 | |
XM_011519752.2 | XP_011518054.1 | 14 - 15 | 2005 | 67 | |
GenBank mRNA | AK297740.1 | - | 13 - 14 | 1478 | 67 |
AK308240.1 | - | 14 - 15 | 1528 | 67 | |
L36531.1 | - | 14 - 15 | 1333 | 67 |