The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005340.6 | NP_005331.1 | 2 - 3 | 360 | 120 |
NR_024611.2 | 2 - 3 | 395 | 120 | ||
GenBank mRNA | AK026557.1 | - | 2 - 3 | 269 | 120 |
BC001287.1 | - | 2 - 3 | 239 | 120 | |
BC007090.1 | - | 2 - 3 | 312 | 120 | |
BF967059.1 | - | 2 - 3 | 501 | 120 | |
BG708765.1 | - | 2 - 3 | 365 | 120 | |
CK002748.1 | - | 2 - 3 | 383 | 120 | |
CN290233.1 | - | 2 - 3 | 212 | 121 | |
CN290239.1 | - | 2 - 3 | 337 | 120 | |
CR457048.1 | - | 2 - 3 | 217 | 120 | |
DB125655.1 | - | 2 - 3 | 347 | 120 | |
U27143.1 | - | 2 - 3 | 260 | 120 | |
U51004.1 | - | 2 - 3 | 324 | 120 |