The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004843.3 | NP_004834.1 | 11 - 12 | 1661 | 67 |
GenBank mRNA | AF106912.1 | - | 11 - 12 | 1653 | 67 |
AF265242.1 | - | 11 - 12 | 1639 | 67 | |
AY358876.1 | - | 11 - 12 | 1639 | 67 | |
BC028003.1 | - | 11 - 12 | 1584 | 67 |