The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004843.3 | NP_004834.1 | 1 - 2 | 236 | 77 |
GenBank mRNA | AF053004.1 | - | 1 - 2 | 241 | 77 |
AF106912.1 | - | 1 - 2 | 228 | 77 | |
AF265242.1 | - | 1 - 2 | 214 | 77 | |
AY358876.1 | - | 1 - 2 | 214 | 77 | |
BC028003.1 | - | 1 - 2 | 159 | 77 |