The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145811.2 | NP_665810.1 | 2 - 3 | 335 | 83 |
GenBank mRNA | AF361351.1 | - | 2 - 3 | 275 | 83 |
AF458898.1 | - | 2 - 3 | 275 | 83 | |
AK290171.1 | - | 3 - 4 | 512 | 83 | |
BC101607.1 | - | 2 - 3 | 333 | 83 | |
BC101633.1 | - | 2 - 3 | 335 | 83 |