Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF26
Entrez Gene ID
7574
Gene Name
zinc finger protein 26
Gene Aliases
HEL-179, KOX20
Chromosome Location
Chr.12: 132986348 - 133027102 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001256280.1 NP_001243209.1 1 - 2 523 83
NM_019591.3 NP_062537.2 1 - 2 523 83
GenBank mRNA BC046206.2 - 1 - 2 424 83
CN268178.1 - 1 - 2 428 83
DA737586.1 - 1 - 2 469 83
EU668356.1 - 1 - 2 401 83

View Full Product Details